Tisk…

4-Hydroxy-3-methoxy-ɑ-methylbenzyl alcohol 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Hydroxy-5-(1-hydroxyethyl)anisole, 4-(1-Hydroxyethyl)-2-methoxyphenol

OR912488-1G OR912488-5G
APOSOR912488-1GEA 2070 CZK
APOSOR912488-1G APOSOR912488-5G
4-Hydroxy-3-methoxy-ɑ-methylbenzyl alcohol 95%
4-Hydroxy-3-methoxy-alpha-methylbenzyl alcohol
Vzorec: C₉H₁₂O₃
MW: 168,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238638
CAS číslo: 2480-86-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR