Tisk…

3-Chloro-4-nitropyridine-1-oxide 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Chloro-4-nitropyridine N-oxide

OR912472-10G OR912472-1G OR912472-25G
APOSOR912472-10GEA 9330 CZK
APOSOR912472-10G APOSOR912472-1G APOSOR912472-25G
3-Chloro-4-nitropyridine-1-oxide 97%
3-Chloro-4-nitropyridine-1-oxide
Vzorec: C₅H₃ClN₂O₃
MW: 174.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00233948
CAS číslo: 76439-45-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR