Tisk…

3-(2-Aminoethyl)pyridine 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-(Pyridin-3-yl)ethanamine, 2-(3-Pyridyl)ethylamine

OR912432-10G OR912432-1G OR912432-25G
APOSOR912432-10GEA 5790 CZK
APOSOR912432-10G APOSOR912432-1G APOSOR912432-25G
3-(2-Aminoethyl)pyridine 98%
3-(2-Aminoethyl)pyridine
Vzorec: C₇H₁₀N₂
Bod varu: 128 °C/24mmHg
Hustota: 1,04 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00191604
CAS číslo: 20173-24-4
UN: 2735
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR