Tisk…

4-Ethylphenetole 95%

Dodavatel: Apollo Scientific

OR912425-1G OR912425-25G OR912425-5G
APOSOR912425-1GEA 1950 CZK
APOSOR912425-1G APOSOR912425-25G APOSOR912425-5G
4-Ethylphenetole 95%
4-Ethylphenetole
Vzorec: C₁₀H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00191432
CAS číslo: 1585-06-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR