Tisk…

3-Chloropyridine N-Oxide 98%

Dodavatel: Apollo Scientific

Synonyms: 3-Chloropyridine 1-oxide

OR912359-5G
APOSOR912359-5GEA 2440 CZK
APOSOR912359-5G
3-Chloropyridine N-Oxide 98%
3-Chloropyridine N-Oxide
Vzorec: C₅H₄ClNO
MW: 129,54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128857
CAS číslo: 1851-22-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR