3,4-Dichloroanisole 97%
Dodavatel: Apollo Scientific
OR912314-25G
APOSOR912314-25GEA
1780
CZK
APOSOR912314-25G
3,4-Dichloroanisole 97%
3,4-Dichloroanisole
Vzorec:
C₇H₆Cl₂O MW: 177,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00070785 CAS číslo: 36404-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...