N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N-Boc-O-benzyl-L-tyrosine, BOC-O-Benzyl-L-tyrosine, Boc-Tyr(Bzl)-OH
OR912300-100G
OR912300-500G
OR912300-25G
APOSOR912300-100GEA
3770
CZK
APOSOR912300-100G
APOSOR912300-500G
APOSOR912300-25G
N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine 98%
N-(tert-Butoxycarbonyl)-O-benzyl-L-tyrosine
Vzorec:
C₂₁H₂₅NO₅ Bod tání: 110…112 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00065597 CAS číslo: 2130-96-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...