Tisk…

Ethyl-2-(2,4,5-trioxo-1-imidazolidinyl)acetate

Dodavatel: Apollo Scientific

OR9123-1G OR9123-5G
APOSOR9123-1GEA 4930 CZK
APOSOR9123-1G APOSOR9123-5G
Ethyl-2-(2,4,5-trioxo-1-imidazolidinyl)acetate
Ethyl-2-(2,4,5-trioxo-1-imidazolidinyl)acetate
Vzorec: C₇H₈N₂O₅
MW: 200,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00665817
CAS číslo: 89694-35-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR