Tisk…

1-(4-Nitrophenyl)imidazole 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(4-Nitrophenyl)-1H-imidazole

OR912274-5G
APOSOR912274-5GEA 1700 CZK
APOSOR912274-5G
1-(4-Nitrophenyl)imidazole 95%
1-(4-Nitrophenyl)imidazole
Vzorec: C₉H₇N₃O₂
MW: 189,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060491
CAS číslo: 2301-25-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR