Tisk…

3-(m-Tolyl)propionic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 3-(3-Methylphenyl)propionic acid, 3-(3-Methylphenyl)propanoic acid

OR912273-5G
APOSOR912273-5GEA 2690 CZK
APOSOR912273-5G
3-(m-Tolyl)propionic acid 97%
3-(m-Tolyl)propionic acid
Vzorec: C₁₀H₁₂O₂
MW: 164,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060326
CAS číslo: 3751-48-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR