trans-4-Butylcyclohexanecarboxylic acid 98%
Dodavatel: Apollo Scientific
OR912272-100G
APOSOR912272-100GEA
1910
CZK
APOSOR912272-100G
trans-4-Butylcyclohexanecarboxylic acid 98%
trans-4-Butylcyclohexanecarboxylic acid
Vzorec:
C₁₁H₂₀O₂ MW: 184,28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00059560 CAS číslo: 38289-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...