Tisk…

2-Aminoimidazole sulphate

Dodavatel: Apollo Scientific

OR912252-100G OR912252-25G
APOSOR912252-100GEA 6450 CZK
APOSOR912252-100G APOSOR912252-25G
2-Aminoimidazole sulphate
2-Aminoimidazole sulphate
Vzorec: C₃H₇N₃O₄S
MW: 181,1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050811
CAS číslo: 36946-29-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR