Tisk…

2-Propoxybenzaldehyde 98%

Dodavatel: Apollo Scientific

Synonyms: 2-n-Propoxybenzaldehyde

OR912242-10G OR912242-1G OR912242-5G
APOSOR912242-10GEA 24600 CZK
APOSOR912242-10G APOSOR912242-1G APOSOR912242-5G
2-Propoxybenzaldehyde 98%
2-Propoxybenzaldehyde
Vzorec: C₁₀H₁₂O₂
MW: 164.2
Storage Temperature: Ambient
MDL Number: MFCD00046150
CAS číslo: 7091-12-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR