Ethyl-3-cyclopentene-1-carboxylate 95%
Dodavatel: Apollo Scientific
OR912181-1G
OR912181-25G
OR912181-5G
APOSOR912181-1GEA
655
CZK
APOSOR912181-1G
APOSOR912181-25G
APOSOR912181-5G
Ethyl-3-cyclopentene-1-carboxylate 95%
Ethyl-3-cyclopentene-1-carboxylate
Vzorec:
C₈H₁₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD06410819 CAS číslo: 21622-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...