4-Pentenoic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
Allylacetic acid, Pent-4-enoic acid
OR912162-100G
OR912162-25G
APOSOR912162-100GEA
1580
CZK
APOSOR912162-100G
APOSOR912162-25G
4-Pentenoic acid 98%
4-Pentenoic acid
Vzorec:
C₅H₈O₂ MW: 100,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004408 CAS číslo: 591-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...