Bis(4-methoxybenzyl)amine 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR912160-1G
OR912160-25G
OR912160-5G
APOSOR912160-1GEA
622
CZK
APOSOR912160-1G
APOSOR912160-25G
APOSOR912160-5G
Bis(4-methoxybenzyl)amine 98%
Bis(4-methoxybenzyl)amine
Vzorec:
C₁₆H₁₉NO₂ MW: 257,333 g/mol Bod tání: 28…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00277836 CAS číslo: 17061-62-0 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...