Tisk…

2-Amino-2'-methyldiphenyl ether 98%

Dodavatel: Apollo Scientific

Synonyms: 2-(o-Tolyloxy)aniline, ['2-Aminophenyl ', 'o', '-Tolyl Ether', '2-(', 'o', '-Tolyloxy)aniline'], 2-Aminophenyl o-Tolyl Ether, 2-Amino-2'-methyldiphenyl Ether

OR912101-100G OR912101-25G OR912101-5G
APOSOR912101-100GEA 15110 CZK
APOSOR912101-100G APOSOR912101-25G APOSOR912101-5G
2-Amino-2'-methyldiphenyl ether 98%
2-Amino-2'-methyldiphenyl ether
Vzorec: C₁₃H₁₃NO
MW: 199.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00025167
CAS číslo: 3840-18-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR