5-Chloro-2,3-dihydroxypyridine 95%
Dodavatel: Apollo Scientific
OR912091-10G
OR912091-5G
APOSOR912091-10GEA
8070
CZK
APOSOR912091-10G
APOSOR912091-5G
5-Chloro-2,3-dihydroxypyridine 95%
5-Chloro-2,3-dihydroxypyridine
Vzorec:
C₅H₄ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00023474 CAS číslo: 53233-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...