Tisk…

5-Chloro-2-methylbenzoxazole 96%

Dodavatel: Apollo Scientific

Synonyms: 5-Chloro-2-methylbenzo[d]oxazole

OR912088-100G OR912088-500G
APOSOR912088-100GEA 2320 CZK
APOSOR912088-100G APOSOR912088-500G
5-Chloro-2-methylbenzoxazole 96%
5-Chloro-2-methylbenzoxazole
Vzorec: C₈H₆ClNO
MW: 167,5900 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022850
CAS číslo: 19219-99-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR