Tisk…
Terebic acid 95%
Dodavatel: Apollo Scientific
Synonyms: Tetrahydro-2,2-dimethyl-5-oxo-3-furoic Acid
OR912078-1G OR912078-250MG
APOSOR912078-1GEA 9330 CZK
APOSOR912078-1G APOSOR912078-250MG
Terebic acid 95%
Terebic acid
Vzorec: C₇H₁₀O₄
MW: 158,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00022485
CAS číslo: 79-91-4