Tisk…

1-Butyl-2-pyrrolidone

Dodavatel: Apollo Scientific
Varování

Synonyms: N-Butyl-2-azacyclopentanone, 1-Butylpyrrolidin-2-one

OR912073-25G OR912073-5G
APOSOR912073-25GEA 3190 CZK
APOSOR912073-25G APOSOR912073-5G
1-Butyl-2-pyrrolidone
1-Butyl-2-pyrrolidone
Vzorec: C₈H₁₅NO
MW: 141,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020876
CAS číslo: 3470-98-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR