Tisk…

3-Methylisonicotinic acid 95%

Dodavatel: Apollo Scientific

Synonyms: 3-Methyl-4-pyridinecarboxylic acid

OR912065-10G OR912065-1G OR912065-5G
APOSOR912065-10GEA 4570 CZK
APOSOR912065-10G APOSOR912065-1G APOSOR912065-5G
3-Methylisonicotinic acid 95%
3-Methylisonicotinic acid
Vzorec: C₇H₇NO₂
MW: 137.1360 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00128870
CAS číslo: 4021-12-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR