4,5,6,7-Tetrahydroisobenzofuran-1,3-dione 97%
Dodavatel: Apollo Scientific
OR912043-100G
OR912043-1KG
OR912043-500G
APOSOR912043-100GEA
4140
CZK
APOSOR912043-100G
APOSOR912043-1KG
APOSOR912043-500G
4,5,6,7-Tetrahydroisobenzofuran-1,3-dione 97%
4,5,6,7-Tetrahydroisobenzofuran-1,3-dione
Vzorec:
C₈H₈O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00005917 CAS číslo: 2426-02-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...