Tisk…

(3-Hydroxy-4-methoxyphenyl)acetic acid 95%

Dodavatel: Apollo Scientific

OR912038-1G OR912038-25G OR912038-5G
APOSOR912038-1GEA 0 CZK
APOSOR912038-1G APOSOR912038-25G APOSOR912038-5G
(3-Hydroxy-4-methoxyphenyl)acetic acid 95%
(3-Hydroxy-4-methoxyphenyl)acetic acid
Vzorec: (C₆H₃(OH)(OCH₃))CH₂COOH
Storage Temperature: Ambient
MDL Number: MFCD00016829
CAS číslo: 1131-94-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR