Tisk…

4-Ethoxyphenylacetonitrile 95%

Dodavatel: Apollo Scientific

OR912015-25G OR912015-5G
APOSOR912015-25GEA 5530 CZK
APOSOR912015-25G APOSOR912015-5G
4-Ethoxyphenylacetonitrile 95%
4-Ethoxyphenylacetonitrile
Vzorec: C₁₀H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00016401
CAS číslo: 6775-77-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR