4-Methyl-1-indanone 95%
Dodavatel: Apollo Scientific
Synonyms:
4-Methyl-2,3-dihydro-1H-inden-1-one
OR912013-5G
APOSOR912013-5GEA
2610
CZK
APOSOR912013-5G
4-Methyl-1-indanone 95%
4-Methyl-1-indanone
Vzorec:
C₁₀H₁₀O MW: 146,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00015759 CAS číslo: 24644-78-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...