Tisk…

(R)-(+)-N-Benzyl-α-methylbenzylamine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (+)-Benzylphenethylamine, (R)-N-Benzyl-1-phenylethanamine, (R)-(+)-N-Benzyl-1-phenylethylamine

OR911995-100G OR911995-25G OR911995-500G
APOSOR911995-100GEA 1330 CZK
APOSOR911995-100G APOSOR911995-25G APOSOR911995-500G
(R)-(+)-N-Benzyl-α-methylbenzylamine 98%
(R)-(+)-N-Benzyl-α-methylbenzylamine
Vzorec: C₁₅H₁₇N
Bod varu: 171 °C/15mmHg
Hustota: 1,01 g/cm³ (25 °C)
Bod vzplanutí: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00015010
CAS číslo: 38235-77-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR