Tisk…

2,4,6-Trimethylbenzyl alcohol 97%

Dodavatel: Apollo Scientific

Synonyms: (2,4,6-Trimethylphenyl)methanol, Mesitylmethanol

OR911974-25G
APOSOR911974-25GEA 5790 CZK
APOSOR911974-25G
2,4,6-Trimethylbenzyl alcohol 97%
2,4,6-Trimethylbenzyl alcohol
Vzorec: C₁₀H₁₄O
MW: 150,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014422
CAS číslo: 4170-90-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR