Tisk…
Ethyl-4-chlorobenzoate 98%
Dodavatel: Apollo Scientific
Synonyms: Ethyl 4-chlorobenzoate, 4-Chlorobenzoic acid ethyl ester
OR911952-100G OR911952-1KG OR911952-500G
APOSOR911952-100GEA 4770 CZK
APOSOR911952-100G APOSOR911952-1KG APOSOR911952-500G
Ethyl-4-chlorobenzoate 98%
Ethyl-4-chlorobenzoate
Vzorec: C₉H₉ClO₂
MW: 184,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013645
CAS číslo: 7335-27-5