Tisk…
Ethyl-3-chlorobenzoate 98%
Dodavatel: Apollo Scientific
Synonyms: 3-Chlorobenzoic acid ethyl ester, Ethyl 3-chlorobenzoate
OR911951-100G
APOSOR911951-100GEA 2440 CZK
APOSOR911951-100G
Ethyl-3-chlorobenzoate 98%
Ethyl-3-chlorobenzoate
Vzorec: C₉H₉ClO₂
MW: 184,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013634
CAS číslo: 1128-76-3