Tisk…

Methyl 3-Hydroxyphenylacetate 97%

Dodavatel: Apollo Scientific

Synonyms: Methyl 2-(3-hydroxyphenyl)acetate, 3-Hydroxyphenylacetic acid methyl ester

OR911917-25G
APOSOR911917-25GEA 4470 CZK
APOSOR911917-25G
Methyl 3-Hydroxyphenylacetate 97%
Methyl 3-Hydroxyphenylacetate
Vzorec: C₉H₁₀O₃
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017660
CAS číslo: 42058-59-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR