4-(2-Hydroxyethyl)oxindole 98%
Dodavatel: Apollo Scientific
Synonyms:
4-(2-Hydroxyethyl)indolin-2-one, 4-(2-Hydroxyethyl)-2-indolinone
OR911911-5G
APOSOR911911-5GEA
2320
CZK
APOSOR911911-5G
4-(2-Hydroxyethyl)oxindole 98%
4-(2-Hydroxyethyl)oxindole
Vzorec:
C₁₀H₁₁NO₂ MW: 177,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07782133 CAS číslo: 139122-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...