Methyl-4-methylthiazole-5-carboxylate 95%
Dodavatel: Apollo Scientific
OR911845-5G
APOSOR911845-5GEA
1870
CZK
APOSOR911845-5G
Methyl-4-methylthiazole-5-carboxylate 95%
Methyl-4-methylthiazole-5-carboxylate
Vzorec:
C₆H₇NO₂S Storage Temperature: Ambient |
CAS číslo:
81569-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...