Tisk…

4-(Methylthio)benzyl alcohol 95%

Dodavatel: Apollo Scientific

Synonyms: 4-(Methylthio)benzyl Alcohol

OR911843-25G OR911843-5G
APOSOR911843-25GEA 5530 CZK
APOSOR911843-25G APOSOR911843-5G
4-(Methylthio)benzyl alcohol 95%
4-(Methylthio)benzyl alcohol
Vzorec: C₈H₁₀OS
Storage Temperature: Ambient
MDL Number: MFCD00009706
CAS číslo: 3446-90-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR