Isoquinolin-N-oxide 95%
Dodavatel: Apollo Scientific
Synonyms:
Isoquinoline N-Oxide
OR911816-100G
OR911816-25G
APOSOR911816-100GEA
11210
CZK
APOSOR911816-100G
APOSOR911816-25G
Isoquinolin-N-oxide 95%
Isoquinolin-N-oxide
Vzorec:
C₉H₇NO Storage Temperature: Ambient |
MDL Number:
MFCD00006903 CAS číslo: 1532-72-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...