Ethyl 2,3-Dicyanopropionate 95%
Dodavatel: Apollo Scientific
OR911785-1G
OR911785-25G
OR911785-100G
APOSOR911785-1GEA
1460
CZK
APOSOR911785-1G
APOSOR911785-25G
APOSOR911785-100G
Ethyl 2,3-Dicyanopropionate 95%
Ethyl 2,3-Dicyanopropionate
Vzorec:
C₇H₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD08061844 CAS číslo: 40497-11-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...