Tisk…

6-Chloroindole-2-carboxylic acid 98%

Dodavatel: Apollo Scientific

OR911762-1G OR911762-25G OR911762-5G
APOSOR911762-1GEA 2610 CZK
APOSOR911762-1G APOSOR911762-25G APOSOR911762-5G
6-Chloroindole-2-carboxylic acid 98%
6-Chloroindole-2-carboxylic acid
Vzorec: C₉H₆ClNO₂
MW: 195,602 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01863157
CAS číslo: 16732-75-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR