Tisk…

4-Pyridylacetonitrile hydrochloride 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(Pyridin-4-yl)acetonitrile hydrochloride

OR911754-1G OR911754-25G OR911754-5G
APOSOR911754-1GEA 1580 CZK
APOSOR911754-1G APOSOR911754-25G APOSOR911754-5G
4-Pyridylacetonitrile hydrochloride 95%
4-Pyridylacetonitrile hydrochloride
Vzorec: C₇H₇ClN₂
MW: 154.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012825
CAS číslo: 92333-25-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR