Tisk…

3-Isochromanone 98%

Dodavatel: Apollo Scientific

Synonyms: 1,4-Dihydro-3H-2-benzopyran-3-one

OR911720-500G
APOSOR911720-500GEA 3270 CZK
APOSOR911720-500G
3-Isochromanone 98%
3-Isochromanone
Vzorec: C₉H₈O₂
MW: 148,16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043005
CAS číslo: 4385-35-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR