Tisk…

Decahydroquinoline 95%

Dodavatel: Apollo Scientific

OR911710-5G
APOSOR911710-5GEA 3190 CZK
APOSOR911710-5G
Decahydroquinoline 95%
Decahydroquinoline
Vzorec: C₉H₁₇N
MW: 139,242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006695
CAS číslo: 2051-28-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR