N-Ethyl-4-hydroxypiperidine 97%
Dodavatel: Apollo Scientific
OR911703-1G
OR911703-5G
APOSOR911703-1GEA
0
CZK
APOSOR911703-1G
APOSOR911703-5G
N-Ethyl-4-hydroxypiperidine 97%
N-Ethyl-4-hydroxypiperidine
Vzorec:
C₇H₁₅NO Storage Temperature: Ambient |
MDL Number:
MFCD00006509 CAS číslo: 3518-83-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...