Tisk…

(S)-(-)-4-Chloro-α-methylbenzylamine 98%

Dodavatel: Apollo Scientific

Synonyms: (S)-1-(4-Chlorophenyl)ethylamine

OR911668-100G OR911668-25G OR911668-5G
APOSOR911668-100GEA 0 CZK
APOSOR911668-100G APOSOR911668-25G APOSOR911668-5G
(S)-(-)-4-Chloro-α-methylbenzylamine 98%
(S)-(-)-4-Chloro-α-methylbenzylamine
Vzorec: C₈H₁₀ClN
MW: 155,63 g/mol
Bod varu: 232 °C/760mmHg
Hustota: 1,115 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00671640
CAS číslo: 4187-56-8
UN: 2810
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR