Tisk…
4-Butylcyclohexanecarboxylic acid (cis and trans mixture) 98%
Dodavatel: Apollo Scientific
Synonyms: 4-Butylcyclohexanecarboxylic acid
OR911645-1G OR911645-25G OR911645-5G
APOSOR911645-1GEA 2900 CZK
APOSOR911645-1G APOSOR911645-25G APOSOR911645-5G
4-Butylcyclohexanecarboxylic acid (cis and trans mixture) 98%
4-Butylcyclohexanecarboxylic acid (cis and trans mixture)
Vzorec: C₁₁H₂₀O₂
MW: 184,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00460757
CAS číslo: 71101-89-8