Methyl 4-Isopropylbenzoate 97%
Dodavatel: Apollo Scientific
Synonyms:
4-Isopropylbenzoic acid methyl ester, Methyl 4-isopropylbenzoate, Methyl Cumate
OR911559-1G
OR911559-25G
OR911559-5G
APOSOR911559-1GEA
1990
CZK
APOSOR911559-1G
APOSOR911559-25G
APOSOR911559-5G
Methyl 4-Isopropylbenzoate 97%
Methyl 4-Isopropylbenzoate
Vzorec:
C₁₁H₁₄O₂ MW: 178,23 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06203666 CAS číslo: 20185-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...