Tisk…

2-Propylpyridine-4-carbothioamide 98%

Dodavatel: Apollo Scientific

Synonyms: 2-Propyl-4-pyridinecarbothioamide, 2-Propylthioisonicotinamide, 2-Propylpyridine-4-carbothioamide, Prothionamide

OR911524-100G OR911524-25G
APOSOR911524-100GEA 6190 CZK
APOSOR911524-100G APOSOR911524-25G
2-Propylpyridine-4-carbothioamide 98%
2-Propylpyridine-4-carbothioamide
Vzorec: C₉H₁₂N₂S
MW: 180,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464119
CAS číslo: 14222-60-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR