α-Methylcinnamic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Methyl-3-phenyl-2-propenoic acid
OR911424-100G
OR911424-25G
APOSOR911424-100GEA
2150
CZK
APOSOR911424-100G
APOSOR911424-25G
α-Methylcinnamic acid 97%
α-Methylcinnamic acid
Vzorec:
C₁₀H₁₀O₂ MW: 162,19 g/mol Bod tání: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002652 CAS číslo: 1199-77-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...