Tisk…

Ethyl-m-anisate 98%

Dodavatel: Apollo Scientific

Synonyms: 3-Anisic acid ethyl ester, Ethyl 3-methoxybenzoate, 3-Methoxybenzoic acid ethyl ester, m-Anisic acid ethyl ester

OR911401-100G
APOSOR911401-100GEA 2780 CZK
APOSOR911401-100G
Ethyl-m-anisate 98%
Ethyl-m-anisate
Vzorec: C₁₀H₁₂O₃
MW: 180,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00015149
CAS číslo: 10259-22-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR