Tisk…
(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid 98%
Dodavatel: Apollo Scientific
Synonyms: (R)-2-(4-Hydroxyphenoxy)propanoic acid
OR911332-500G
APOSOR911332-500GEA 2860 CZK
APOSOR911332-500G
(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid 98%
(R)-(+)-2-(4-Hydroxyphenoxy)propionic acid
Vzorec: C₉H₁₀O₄
MW: 182,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00274088
CAS číslo: 94050-90-5