Tisk…

2-Ethylbenzofuran 98%

Dodavatel: Apollo Scientific

OR911327-100G
APOSOR911327-100GEA 3710 CZK
APOSOR911327-100G
2-Ethylbenzofuran 98%
2-Ethylbenzofuran
Vzorec: C₁₀H₁₀O
MW: 146,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00047287
CAS číslo: 3131-63-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR