Tisk…

4-Amino-2-chloro-N,N-dimethylbenzamide 98%

Dodavatel: Apollo Scientific

OR911255-10G OR911255-1G OR911255-5G
APOSOR911255-10GEA 26450 CZK
APOSOR911255-10G APOSOR911255-1G APOSOR911255-5G
4-Amino-2-chloro-N,N-dimethylbenzamide 98%
4-Amino-2-chloro-N,N-dimethylbenzamide
Vzorec: C₉H₁₁ClN₂O
Storage Temperature: Ambient
CAS číslo: 98995-06-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR